Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Baz1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011714.2 | NP_035844.2 | 11 - 12 | 3402 | 116 | |
GenBank mRNA | AK037737.1 | - | 5 - 6 | 610 | 116 | |
BC141399.1 | - | 11 - 12 | 3305 | 9056012 | 116 | |
BC171975.1 | - | 11 - 12 | 3304 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |