Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021535.4 | NP_067510.3 | 1 - 2 | 108 | 75 | |
GenBank mRNA | AB044414.1 | - | 1 - 2 | 37 | 75 | |
AK003493.1 | - | 1 - 2 | 95 | 75 | ||
AK011140.1 | - | 1 - 2 | 107 | 75 | ||
AK019174.1 | - | 1 - 2 | 67 | 75 | ||
AK145982.1 | - | 1 - 2 | 107 | 75 | ||
AK159635.1 | - | 1 - 2 | 95 | 75 | ||
AK168813.1 | - | 1 - 2 | 102 | 75 | ||
BC020034.1 | - | 1 - 2 | 50 | 75 | ||
BC057446.1 | - | 1 - 2 | 69 | 3155760 | 75 | |
BY036103.1 | - | 1 - 2 | 110 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |