Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010084.2 | NP_034214.2 | 5 - 6 | 395 | 80 | |
GenBank mRNA | AF167405.1 | - | 5 - 6 | 393 | 80 | |
AK076633.1 | - | 5 - 6 | 396 | 80 | ||
BC051136.1 | - | 5 - 6 | 372 | 6708158 | 80 | |
BC144841.1 | - | 5 - 6 | 368 | 80 | ||
BC145751.1 | - | 5 - 6 | 368 | 40131054 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |