Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tacc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177089.5 | NP_796063.3 | 5 - 6 | 1825 | 68 | |
NM_199323.3 | NP_955355.1 | 3 - 4 | 601 | 68 | ||
GenBank mRNA | AK046384.1 | - | 6 - 7 | 1455 | 68 | |
AK138513.1 | - | 3 - 4 | 476 | 68 | ||
AY177412.1 | - | 3 - 4 | 509 | 68 | ||
AY177413.1 | - | 5 - 6 | 1727 | 68 | ||
BC125389.1 | - | 3 - 4 | 411 | 68 | ||
BC145709.1 | - | 3 - 4 | 411 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |