Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hlcs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139145.4 | NP_631884.1 | 6 - 7 | 1687 | 106 | |
GenBank mRNA | AB066227.1 | - | 5 - 6 | 1547 | 106 | |
AK052424.1 | - | 7 - 8 | 1983 | 106 | ||
AK158212.1 | - | 7 - 8 | 1804 | 106 | ||
AK168925.1 | - | 3 - 4 | 582 | 106 | ||
BC050090.1 | - | 6 - 7 | 1688 | 30012868 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |