Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryzl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026994.1 | NP_081270.1 | 7 - 8 | 785 | 91 | |
NM_133679.2 | NP_598440.1 | 8 - 9 | 830 | 91 | ||
GenBank mRNA | AK010433.1 | - | 8 - 9 | 634 | 91 | |
AK145827.1 | - | 8 - 9 | 654 | 91 | ||
BC010479.1 | - | 8 - 9 | 627 | 3968727 | 91 | |
BC019387.1 | - | 7 - 8 | 594 | 4164847 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |