Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ablim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177696.1 | NP_001171167.1 | 4 - 5 | 619 | 93 | |
NM_001177697.1 | NP_001171168.1 | 4 - 5 | 619 | 93 | ||
NM_001177698.1 | NP_001171169.1 | 4 - 5 | 619 | 93 | ||
NM_001177699.1 | NP_001171170.1 | 4 - 5 | 619 | 93 | ||
NM_001177700.1 | NP_001171171.1 | 4 - 5 | 619 | 93 | ||
NM_177678.7 | NP_808346.3 | 4 - 5 | 619 | 93 | ||
GenBank mRNA | AJ748602.1 | - | 4 - 5 | 575 | 93 | |
AJ748603.1 | - | 4 - 5 | 575 | 93 | ||
AK046243.1 | - | 4 - 5 | 570 | 93 | ||
AK046879.1 | - | 4 - 5 | 568 | 93 | ||
AK082707.1 | - | 4 - 5 | 569 | 93 | ||
AK154494.1 | - | 4 - 5 | 621 | 93 | ||
AY274116.1 | - | 4 - 5 | 546 | 93 | ||
BC141125.1 | - | 4 - 5 | 479 | 9055737 | 93 | |
DQ413175.1 | - | 3 - 4 | 448 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |