Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gata5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008093.2 | NP_032119.2 | 2 - 3 | 1032 | 66 | |
GenBank mRNA | AK132020.1 | - | 2 - 3 | 1053 | 66 | |
AK142213.1 | - | 2 - 3 | 1034 | 66 | ||
BC105653.1 | - | 3 - 4 | 745 | 40046623 | 66 | |
BC105654.1 | - | 3 - 4 | 806 | 40046622 | 66 | |
BC105655.1 | - | 3 - 4 | 655 | 40046621 | 66 | |
BC105656.1 | - | 3 - 4 | 311 | 66 | ||
U84725.1 | - | 2 - 3 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |