Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2cd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174847.2 | NP_777272.2 | 12 - 13 | 1867 | 78 | |
GenBank mRNA | AK077731.1 | - | 12 - 13 | 1828 | 78 | |
AK080956.1 | - | 12 - 13 | 1811 | 78 | ||
AK086520.1 | - | 12 - 13 | 1870 | 78 | ||
AK143875.1 | - | 12 - 13 | 1841 | 78 | ||
AK169902.1 | - | 12 - 13 | 1803 | 78 | ||
AK196045.1 | - | 3 - 4 | 302 | 78 | ||
AK220454.1 | - | 12 - 13 | 1852 | 78 | ||
BC038064.1 | - | 2 - 3 | 299 | 78 | ||
BC043688.1 | - | 12 - 13 | 1828 | 5136661 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |