Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pear1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032413.1 | NP_001027585.1 | 8 - 9 | 972 | 89 | |
NM_001032414.1 | NP_001027586.1 | 7 - 8 | 902 | 89 | ||
NM_001289600.1 | NP_001276529.1 | 7 - 8 | 874 | 89 | ||
NM_001289601.1 | NP_001276530.1 | 9 - 10 | 1115 | 89 | ||
NM_028460.2 | NP_082736.1 | 9 - 10 | 1106 | 89 | ||
NM_152799.2 | NP_690012.2 | 9 - 10 | 1071 | 89 | ||
GenBank mRNA | AF440279.1 | - | 9 - 10 | 1096 | 89 | |
AF444274.1 | - | 7 - 8 | 896 | 89 | ||
AF461685.1 | - | 6 - 7 | 829 | 89 | ||
AK053551.1 | - | 8 - 9 | 963 | 89 | ||
AK141082.1 | - | 869 | 89 | |||
AK161049.1 | - | 9 - 10 | 1112 | 89 | ||
BC042490.1 | - | 8 - 9 | 921 | 4953584 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |