Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Veph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145820.3 | NP_665819.2 | 10 - 11 | 2256 | 80 | |
GenBank mRNA | AB085187.1 | - | 10 - 11 | 2238 | 80 | |
AB085189.1 | - | 2 - 3 | 301 | 80 | ||
AK013398.1 | - | 2 - 3 | 316 | 80 | ||
BC075649.1 | - | 10 - 11 | 2256 | 80 | ||
BC094339.1 | - | 2 - 3 | 269 | 80 | ||
BC128271.1 | - | 9 - 10 | 1963 | 40059997 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |