Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039373.5 | NP_001034462.1 | 2 - 3 | 724 | 63 | |
GenBank mRNA | U32332.1 | - | 2 - 3 | 683 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |