Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh18a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019698.2 | NP_062672.2 | 2 - 3 | 223 | 68 | |
NM_153554.2 | NP_705782.2 | 2 - 3 | 223 | 68 | ||
GenBank mRNA | AF056573.1 | - | 2 - 3 | 128 | 68 | |
AF056574.1 | - | 2 - 3 | 128 | 68 | ||
AK134148.1 | - | 2 - 3 | 183 | 68 | ||
AK147711.1 | - | 2 - 3 | 143 | 68 | ||
AK151072.1 | - | 2 - 3 | 145 | 68 | ||
AK153946.1 | - | 2 - 3 | 144 | 68 | ||
AK159610.1 | - | 2 - 3 | 145 | 68 | ||
AK166459.1 | - | 2 - 3 | 108 | 68 | ||
AK166494.1 | - | 2 - 3 | 108 | 68 | ||
AK168905.1 | - | 2 - 3 | 143 | 68 | ||
BB839822.1 | - | 2 - 3 | 220 | 68 | ||
BC033427.1 | - | 2 - 3 | 154 | 5009115 | 68 | |
BC037699.1 | - | 2 - 3 | 102 | 4188900 | 68 | |
BY126398.1 | - | 2 - 3 | 223 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |