Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Muc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013605.2 | NP_038633.1 | 5 - 6 | 1738 | 77 | |
GenBank mRNA | BC005441.1 | - | 5 - 6 | 1709 | 3601782 | 77 |
BG079301.2 | - | 4 - 5 | 367 | 77 | ||
M84683.1 | - | 5 - 6 | 1688 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |