Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scamp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011886.2 | NP_036016.2 | 2 - 3 | 251 | 88 | |
GenBank mRNA | AF005036.1 | - | 2 - 3 | 158 | 88 | |
AF295403.1 | - | 2 - 3 | 156 | 88 | ||
AK146204.1 | - | 2 - 3 | 253 | 88 | ||
AK160549.1 | - | 2 - 3 | 236 | 88 | ||
AK170115.1 | - | 2 - 3 | 218 | 88 | ||
BY170046.1 | - | 2 - 3 | 251 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |