Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nop14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029278.2 | NP_083554.2 | 12 - 13 | 1842 | 78 | |
GenBank mRNA | AK148605.1 | - | 12 - 13 | 1800 | 78 | |
AK161728.1 | - | 12 - 13 | 1780 | 78 | ||
BC024998.1 | - | 12 - 13 | 1763 | 5012070 | 78 | |
BC043043.1 | - | 12 - 13 | 1785 | 5701430 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |