Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2136776 |
AK033498.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2136776 |
BC024361.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2136776 |
NM_019878.4 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
756971 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
469345 |
AK005079.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
469345 |
AK014648.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
469345 |
AK168847.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
469345 |
BC003849.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
469345 |
NM_023248.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
212364 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
828026 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
119143 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
150383 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
706052 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
931095 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
712529 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
837294 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
476739 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
128098 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P130671 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P178447 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P168618 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2136776 |
AK033498.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2136776 |
BC024361.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2136776 |
NM_019878.4 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P206551 |
AF022894.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P206551 |
AK033498.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P206551 |
BC024361.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P206551 |
NM_019878.4 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P644613 |
AK033498.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P644613 |
NM_019878.4 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23868250 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23868248 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23868255 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23868254 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23868267 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23868269 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23868262 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23868266 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23868257 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23868260 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23868249 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23868253 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23868264 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23868268 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P178447 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05644986 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P130671 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P168618 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P206551 |
AF022894.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P206551 |
AK033498.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P206551 |
BC024361.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P206551 |
NM_019878.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P644613 |
AK033498.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P644613 |
NM_019878.4 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P132831 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P116051 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P168618 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P130671 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P178447 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04800232 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04800233 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04161081 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04800231 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868154 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868249 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868153 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868280 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868164 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868115 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868218 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868131 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868114 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868175 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868146 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868168 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868220 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868118 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868143 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868270 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868181 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868236 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868194 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868224 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868238 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868173 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868226 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868170 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868198 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868273 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868117 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868129 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868144 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868167 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868190 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868185 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868214 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868235 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868171 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868196 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868172 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868212 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868136 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868255 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868124 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868254 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868149 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868268 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868264 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868156 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868278 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868242 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868271 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868213 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868216 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868187 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868126 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868158 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868253 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868260 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868276 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868141 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868119 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868183 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868244 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868262 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868120 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868162 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868229 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868266 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868176 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868130 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868150 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868169 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868121 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868128 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868231 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868179 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868215 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868222 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868239 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868199 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868246 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868192 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868241 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868138 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868201 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868274 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868177 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868217 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23868139 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04241708 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868220 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868259 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04115942 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04603748 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868248 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868257 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868262 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868213 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868246 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868263 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868231 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868217 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868226 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868239 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868236 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868253 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868266 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868210 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868268 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868242 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868206 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868224 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868260 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04278558 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868222 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868235 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04115947 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868244 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868250 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868212 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868258 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868229 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04276248 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868241 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04976473 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868264 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868216 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868249 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23868237 |
- |
- |