Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127382.2 | NP_001120854.1 | 5 - 6 | 1632 | 54 | |
NM_139065.3 | NP_620704.1 | 4 - 5 | 1390 | 54 | ||
NM_178446.4 | NP_848541.2 | 4 - 5 | 1832 | 54 | ||
GenBank mRNA | AK035620.1 | - | 4 - 5 | 1783 | 54 | |
AK040440.1 | - | 1 - 2 | 874 | 54 | ||
AK085733.1 | - | 4 - 5 | 1824 | 54 | ||
AK170082.1 | - | 5 - 6 | 1587 | 54 | ||
BC013481.1 | - | 4 - 5 | 1402 | 4240441 | 54 | |
BC034195.1 | - | 3 - 4 | 1338 | 4984936 | 54 | |
BC112902.1 | - | 4 - 5 | 1832 | 4978873 | 54 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |