Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dthd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170705.1 | NP_001164176.1 | 5 - 6 | 1482 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |