Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083328.1 | NP_001076797.1 | 7 - 8 | 898 | 77 | |
NM_001083329.1 | NP_001076798.1 | 6 - 7 | 804 | 77 | ||
NM_028422.1 | NP_082698.1 | 7 - 8 | 943 | 77 | ||
NM_181424.3 | NP_852089.2 | 8 - 9 | 1037 | 77 | ||
GenBank mRNA | AK007444.1 | - | 5 - 6 | 889 | 77 | |
AK040689.1 | - | 7 - 8 | 1427 | 77 | ||
AK183355.1 | - | 1 - 2 | 45 | 77 | ||
BC028465.1 | - | 2 - 3 | 339 | 77 | ||
DT929445.1 | - | 4 - 5 | 521 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |