Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uckl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026765.3 | NP_081041.2 | 2 - 3 | 311 | 73 | |
GenBank mRNA | AK144803.1 | - | 2 - 3 | 317 | 73 | |
BC016535.1 | - | 2 - 3 | 311 | 2655873 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |