Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igsf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177886.1 | NP_001171357.1 | 6 - 7 | 953 | 87 | |
NM_001177887.1 | NP_001171358.1 | 6 - 7 | 957 | 87 | ||
NM_028078.3 | NP_082354.1 | 5 - 6 | 650 | 87 | ||
GenBank mRNA | AF537215.1 | - | 6 - 7 | 891 | 87 | |
AK008060.1 | - | 5 - 6 | 621 | 87 | ||
AK078934.1 | - | 5 - 6 | 621 | 87 | ||
AK086973.1 | - | 6 - 7 | 955 | 87 | ||
BC004806.1 | - | 5 - 6 | 603 | 3584645 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |