Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161373.1 | NP_001154845.1 | 6 - 7 | 1196 | 85 | |
NM_001161374.1 | NP_001154846.1 | 6 - 7 | 1196 | 85 | ||
NM_007977.2 | NP_032003.2 | 6 - 7 | 1196 | 85 | ||
GenBank mRNA | AJ627040.1 | - | 5 - 6 | 616 | 85 | |
AK051579.1 | - | 6 - 7 | 3472 | 85 | ||
BC138347.1 | - | 6 - 7 | 1077 | 8861367 | 85 | |
BC145245.1 | - | 6 - 7 | 1077 | 9053797 | 85 | |
L05573.1 | - | 6 - 7 | 1194 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |