Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cabp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160252.1 | NP_001153724.1 | 4 - 5 | 379 | 71 | |
NM_001160253.1 | NP_001153725.1 | 4 - 5 | 661 | 71 | ||
NM_013878.2 | NP_038906.2 | 5 - 6 | 538 | 71 | ||
GenBank mRNA | AF169156.1 | - | 4 - 5 | 407 | 71 | |
AF169157.1 | - | 5 - 6 | 566 | 71 | ||
AK007488.1 | - | 4 - 5 | 662 | 71 | ||
BC107029.2 | - | 5 - 6 | 538 | 40050356 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |