Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF012709.1 | - | 3 - 4 | 398 | 118 | |
AK010718.1 | - | 3 - 4 | 360 | 118 | ||
AK011399.1 | - | 3 - 4 | 343 | 118 | ||
AK040313.1 | - | 4 - 5 | 525 | 118 | ||
AK041138.1 | - | 4 - 5 | 518 | 118 | ||
AK053763.1 | - | 3 - 4 | 367 | 118 | ||
AK089033.1 | - | 3 - 4 | 371 | 118 | ||
BC011038.1 | - | 3 - 4 | 378 | 4018429 | 118 | |
BC012280.1 | - | 5 - 6 | 471 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |