Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172988.4 | NP_766576.1 | 2 - 3 | 250 | 67 | |
NR_104378.1 | 2 - 3 | 250 | 67 | |||
NR_104379.1 | 2 - 3 | 250 | 67 | |||
NR_104380.1 | 2 - 3 | 250 | 67 | |||
GenBank mRNA | AK028786.1 | - | 2 - 3 | 234 | 67 | |
AK032391.1 | - | 2 - 3 | 229 | 67 | ||
AK051429.1 | - | 2 - 3 | 201 | 67 | ||
AK083661.1 | - | 2 - 3 | 224 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |