Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081397.1 | NP_001074866.1 | 6 - 7 | 1035 | 93 | |
GenBank mRNA | AB429291.1 | - | 5 - 6 | 709 | 93 | |
AK082350.1 | - | 6 - 7 | 771 | 93 | ||
BC072580.1 | - | 6 - 7 | 1047 | 93 | ||
BC151049.1 | - | 6 - 7 | 989 | 9087962 | 93 | |
BC151051.1 | - | 6 - 7 | 990 | 9087964 | 93 | |
BC157966.1 | - | 6 - 7 | 990 | 9007290 | 93 | |
BC172122.1 | - | 6 - 7 | 990 | 9054408 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |