Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800001402.mm.1 |
NM_029116.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001402.mm.1 |
NM_029116.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001402.mm.1 |
TC0800000150.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800001401.mm.1 |
NM_029116.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001401.mm.1 |
NM_029116.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001401.mm.1 |
TC0800000150.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800001398.mm.1 |
NM_029116.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001398.mm.1 |
NM_029116.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001398.mm.1 |
TC0800000150.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800001403.mm.1 |
NM_029116.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001403.mm.1 |
NM_029116.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001403.mm.1 |
TC0800000150.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800001404.mm.1 |
NM_029116.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001404.mm.1 |
NM_029116.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001404.mm.1 |
TC0800000150.mm.1 |
MTA TC |