Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs3st1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010474.2 | NP_034604.1 | 1 - 2 | 210 | 98 | |
GenBank mRNA | AK087753.1 | - | 1 - 2 | 189 | 98 | |
BC009133.1 | - | 1 - 2 | 172 | 3155049 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |