Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aspdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026690.1 | NP_080966.1 | 5 - 6 | 704 | 69 | |
GenBank mRNA | AK002587.1 | - | 5 - 6 | 704 | 69 | |
BC019451.1 | - | 5 - 6 | 670 | 4194890 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |