Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dntt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009345.2 | NP_033371.2 | 10 - 11 | 1611 | 65 | |
GenBank mRNA | AF316014.1 | - | 10 - 11 | 1464 | 65 | |
AK087978.1 | - | 10 - 11 | 1612 | 65 | ||
AK088709.1 | - | 10 - 11 | 1612 | 65 | ||
X04123.1 | - | 10 - 11 | 1495 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |