Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500016704.mm.1 |
TC0500001808.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016704.mm.1 |
NM_028291.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016704.mm.1 |
NM_028291.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016716.mm.1 |
TC0500001808.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016716.mm.1 |
NM_028291.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016716.mm.1 |
NM_028291.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016721.mm.1 |
TC0500001808.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016721.mm.1 |
NM_028291.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016721.mm.1 |
NM_028291.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016712.mm.1 |
TC0500001808.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016696.mm.1 |
TC0500001808.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016696.mm.1 |
NM_028291.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016696.mm.1 |
NM_028291.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016703.mm.1 |
TC0500001808.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016703.mm.1 |
NM_028291.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016703.mm.1 |
NM_028291.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016720.mm.1 |
TC0500001808.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016720.mm.1 |
NM_028291.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016720.mm.1 |
NM_028291.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016725.mm.1 |
TC0500001808.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016725.mm.1 |
NM_028291.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016725.mm.1 |
NM_028291.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016730.mm.1 |
TC0500001808.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016730.mm.1 |
NM_028291.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016730.mm.1 |
NM_028291.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016695.mm.1 |
TC0500001808.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016695.mm.1 |
NM_028291.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016695.mm.1 |
NM_028291.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016699.mm.1 |
TC0500001808.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016699.mm.1 |
NM_028291.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016699.mm.1 |
NM_028291.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016722.mm.1 |
TC0500001808.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016722.mm.1 |
NM_028291.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016722.mm.1 |
NM_028291.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016728.mm.1 |
TC0500001808.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016728.mm.1 |
NM_028291.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016728.mm.1 |
NM_028291.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016718.mm.1 |
TC0500001808.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016718.mm.1 |
NM_028291.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016718.mm.1 |
NM_028291.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016719.mm.1 |
TC0500001808.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016719.mm.1 |
NM_028291.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016719.mm.1 |
NM_028291.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016709.mm.1 |
TC0500001808.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016709.mm.1 |
NM_028291.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016709.mm.1 |
NM_028291.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016715.mm.1 |
TC0500001808.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016715.mm.1 |
NM_028291.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016715.mm.1 |
NM_028291.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016706.mm.1 |
TC0500001808.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016706.mm.1 |
NM_028291.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016706.mm.1 |
NM_028291.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016729.mm.1 |
TC0500001808.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016729.mm.1 |
NM_028291.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016729.mm.1 |
NM_028291.4 |
Refseq |