Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500016797.mm.1 |
TC0500001821.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016797.mm.1 |
NM_029920.7.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016797.mm.1 |
NM_029920.7 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016787.mm.1 |
TC0500001821.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016787.mm.1 |
NM_029920.7.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016787.mm.1 |
NM_029920.7 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016790.mm.1 |
TC0500001821.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016790.mm.1 |
NM_029920.7.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016790.mm.1 |
NM_029920.7 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016795.mm.1 |
TC0500001821.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016795.mm.1 |
NM_029920.7.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016795.mm.1 |
NM_029920.7 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016788.mm.1 |
TC0500001821.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016788.mm.1 |
NM_029920.7.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016788.mm.1 |
NM_029920.7 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016791.mm.1 |
TC0500001821.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016791.mm.1 |
NM_029920.7.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016791.mm.1 |
NM_029920.7 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016793.mm.1 |
TC0500001821.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016793.mm.1 |
NM_029920.7.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016793.mm.1 |
NM_029920.7 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016801.mm.1 |
TC0500001821.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016801.mm.1 |
NM_029920.7.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016801.mm.1 |
NM_029920.7 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016789.mm.1 |
TC0500001821.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016789.mm.1 |
NM_029920.7.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016789.mm.1 |
NM_029920.7 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016796.mm.1 |
TC0500001821.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016796.mm.1 |
NM_029920.7.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016796.mm.1 |
NM_029920.7 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016798.mm.1 |
TC0500001821.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016798.mm.1 |
NM_029920.7.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016798.mm.1 |
NM_029920.7 |
Refseq |