Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277149.1 | NP_001264078.1 | 36 - 37 | 8492 | 84 | |
GenBank mRNA | BC034239.1 | - | 5 - 6 | 1022 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |