Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007592.3 | NP_031618.2 | 3 - 4 | 502 | 64 | |
GenBank mRNA | AK004896.1 | - | 3 - 4 | 503 | 64 | |
AK004923.1 | - | 3 - 4 | 503 | 64 | ||
BC010773.1 | - | 3 - 4 | 478 | 4216498 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |