Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178387.3 | NP_848474.2 | 4 - 5 | 624 | 77 | |
GenBank mRNA | BC120503.1 | - | 1 - 2 | 76 | 77 | |
BC120505.1 | - | 1 - 2 | 76 | 77 | ||
BC144721.1 | - | 1 - 2 | 76 | 77 | ||
BC144723.1 | - | 1 - 2 | 76 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |