Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncoa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033988.2 | NP_001029160.1 | 5 - 6 | 616 | 66 | |
NM_001284319.1 | NP_001271248.1 | 5 - 6 | 608 | 66 | ||
NM_019744.4 | NP_062718.2 | 5 - 6 | 526 | 66 | ||
GenBank mRNA | AF159461.1 | - | 5 - 6 | 523 | 66 | |
AK014479.1 | - | 5 - 6 | 524 | 66 | ||
AK032951.1 | - | 3 - 4 | 282 | 66 | ||
AK132981.1 | - | 66 | ||||
AK143965.1 | - | 5 - 6 | 521 | 66 | ||
AK145070.1 | - | 5 - 6 | 577 | 66 | ||
AK145945.1 | - | 5 - 6 | 569 | 66 | ||
AK145954.1 | - | 5 - 6 | 66 | |||
AK146384.1 | - | 5 - 6 | 527 | 66 | ||
AK146498.1 | - | 5 - 6 | 566 | 66 | ||
AK146598.1 | - | 5 - 6 | 550 | 66 | ||
AK146646.1 | - | 5 - 6 | 521 | 66 | ||
AK147036.1 | - | 5 - 6 | 524 | 66 | ||
AK147125.1 | - | 5 - 6 | 568 | 66 | ||
AK156999.1 | - | 5 - 6 | 607 | 66 | ||
BC031528.1 | - | 5 - 6 | 567 | 4008986 | 66 | |
BC085086.1 | - | 5 - 6 | 512 | 6335192 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |