Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncapg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019438.1 | NP_062311.1 | 18 - 19 | 2859 | 121 | |
GenBank mRNA | AK136254.1 | - | 13 - 14 | 1845 | 121 | |
BC137834.1 | - | 11 - 12 | 2077 | 8860853 | 121 | |
BC144960.1 | - | 11 - 12 | 2077 | 9053508 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |