Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gucy1b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161796.1 | NP_001155268.1 | 11 - 12 | 1684 | 80 | |
NM_017469.4 | NP_059497.1 | 11 - 12 | 1684 | 80 | ||
GenBank mRNA | AF020339.1 | - | 11 - 12 | 1658 | 80 | |
AF297083.1 | - | 11 - 12 | 1658 | 80 | ||
AK139406.1 | - | 11 - 12 | 1647 | 80 | ||
AK147128.1 | - | 11 - 12 | 1620 | 80 | ||
BC050945.1 | - | 11 - 12 | 1662 | 6822142 | 80 | |
BC056995.1 | - | 11 - 12 | 1662 | 6822142 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |