Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eef1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007906.2 | NP_031932.1 | 1 - 2 | 290 | 129 | |
GenBank mRNA | BC018235.1 | - | 1 - 2 | 290 | 4504385 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |