Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147201.2 | NP_671734.1 | 3 - 4 | 434 | 160 | |
GenBank mRNA | AF302138.1 | - | 3 - 4 | 433 | 160 | |
AK046142.1 | - | 3 - 4 | 428 | 160 | ||
AK077440.1 | - | 3 - 4 | 470 | 160 | ||
AK170501.1 | - | 3 - 4 | 450 | 160 | ||
BC004756.1 | - | 3 - 4 | 404 | 3154089 | 160 | |
BC018463.1 | - | 3 - 4 | 408 | 3598438 | 160 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |