Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_003508.1 | 9 - 10 | 1223 | 77 | ||
GenBank mRNA | AK147949.1 | - | 1843 | 77 | ||
BC007127.1 | - | 9 - 10 | 1206 | 3591798 | 77 | |
J03368.1 | - | 9 - 10 | 1221 | 77 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
family with sequence similarity 3, member B |
Fam3b |
2-21, 9030624C24Rik, D16Jhu19e, ORF9, Pander |
-46.5kb | Anti-sense | Chr. 16 - 97471086 - 97504936 on NCBI Build GRCm38 |
Downstream |
transmembrane protease, serine 2 |
Tmprss2 | D16Ertd61e | 12.6kb | Anti-sense | Chr. 16 - 97564682 - 97659538 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |