Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162947.1 | NP_001156419.1 | 4 - 5 | 434 | 150 | |
NM_011848.4 | NP_035978.2 | 5 - 6 | 492 | 150 | ||
GenBank mRNA | AF093416.1 | - | 4 - 5 | 439 | 150 | |
AF099066.1 | - | 5 - 6 | 461 | 150 | ||
BU058938.1 | - | 5 - 6 | 492 | 150 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |