Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcrl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183222.3 | NP_899045.3 | 3 - 4 | 453 | 64 | |
GenBank mRNA | AY158090.1 | - | 3 - 4 | 453 | 64 | |
AY506558.1 | - | 3 - 4 | 462 | 64 | ||
BC137997.1 | - | 3 - 4 | 390 | 8861016 | 64 | |
BC137998.1 | - | 3 - 4 | 390 | 8861017 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |