Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027493.3 | NP_081769.2 | 8 - 9 | 1191 | 84 | |
GenBank mRNA | AK017815.1 | - | 8 - 9 | 1142 | 84 | |
BC027281.1 | - | 8 - 9 | 1097 | 3590945 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |