Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500023561.mm.1 |
TC0500002675.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500023561.mm.1 |
NM_007683.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023569.mm.1 |
TC0500002675.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500023569.mm.1 |
NM_007683.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023570.mm.1 |
TC0500002675.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500023570.mm.1 |
NM_007683.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023576.mm.1 |
TC0500002675.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500023576.mm.1 |
NM_007683.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023562.mm.1 |
TC0500002675.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500023562.mm.1 |
NM_007683.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023563.mm.1 |
TC0500002675.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500023563.mm.1 |
NM_007683.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023573.mm.1 |
TC0500002675.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500023573.mm.1 |
NM_007683.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023575.mm.1 |
TC0500002675.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500023575.mm.1 |
NM_007683.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023568.mm.1 |
TC0500002675.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500023568.mm.1 |
NM_007683.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023574.mm.1 |
TC0500002675.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500023574.mm.1 |
NM_007683.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023566.mm.1 |
TC0500002675.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500023566.mm.1 |
NM_007683.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023572.mm.1 |
TC0500002675.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500023572.mm.1 |
NM_007683.4 |
Refseq |