Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007529.2 | NP_031555.2 | 10 - 11 | 2413 | 60 | |
GenBank mRNA | AK144405.1 | - | 10 - 11 | 2214 | 60 | |
AK163646.1 | - | 3 - 4 | 474 | 60 | ||
BC052032.1 | - | 10 - 11 | 2208 | 5695027 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |