Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pibf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029320.3 | NP_083596.2 | 14 - 15 | 2563 | 144 | |
GenBank mRNA | AK006060.1 | - | 5 - 6 | 537 | 144 | |
AK017117.1 | - | 5 - 6 | 629 | 144 | ||
AK149007.1 | - | 9 - 10 | 1116 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |