Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028882.4 | NP_083158.3 | 12 - 13 | 1959 | 63 | |
GenBank mRNA | AK028900.1 | - | 12 - 13 | 1741 | 63 | |
AK031704.1 | - | 12 - 13 | 1571 | 63 | ||
AK052671.1 | - | 12 - 13 | 1960 | 63 | ||
AK153839.1 | - | 12 - 13 | 1620 | 63 | ||
BC138131.1 | - | 12 - 13 | 1502 | 8861150 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |