Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081122.1 | NP_001074591.1 | 8 - 9 | 868 | 126 | |
NM_001301689.1 | NP_001288618.1 | 8 - 9 | 868 | 126 | ||
GenBank mRNA | AK076549.1 | - | 8 - 9 | 869 | 126 | |
KC211185.1 | - | 8 - 9 | 725 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |